Assay Details
Gene Symbol: Cry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198750.2 | NP_942045.2 | 6 - 7 | 1351 | 101 | |
XM_008765225.1 | XP_008763447.1 | 6 - 7 | 1375 | 101 | ||
GenBank mRNA | AF545855.1 | - | 6 - 7 | 915 | 101 | |
BC107677.1 | - | 6 - 7 | 1351 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
