Assay Details
Gene Symbol: Fbxo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012222.1 | NP_001012222.1 | 4 - 5 | 815 | 91 | |
XM_006241166.2 | XP_006241228.1 | 4 - 5 | 687 | 91 | ||
XM_006241167.2 | XP_006241229.1 | 4 - 5 | 874 | 91 | ||
XM_006241168.2 | XP_006241230.1 | 4 - 5 | 886 | 91 | ||
XM_008765259.1 | XP_008763481.1 | 4 - 5 | 747 | 91 | ||
GenBank mRNA | BC079382.1 | - | 4 - 5 | 815 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
