Assay Details
Gene Symbol: Glt8d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765296.1 | XP_008763518.1 | 1 - 2 | 643 | 77 | |
XM_008765297.1 | XP_008763519.1 | 1 - 2 | 642 | 77 | ||
XM_008765298.1 | XP_008763520.1 | 1 - 2 | 642 | 77 | ||
XM_008765299.1 | XP_008763521.1 | 1 - 2 | 642 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765296.1, XM_008765297.1, XM_008765298.1, XM_008765299.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
