Assay Details
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082478.1 | NP_001075947.1 | 1 - 2 | 336 | 102 | |
NM_001082479.1 | NP_001075948.1 | 1 - 2 | 336 | 102 | ||
XM_006241191.2 | XP_006241253.1 | 2 - 3 | 180 | 102 | ||
XM_006241192.1 | XP_006241254.1 | 2 - 3 | 94 | 102 | ||
XM_006241193.2 | XP_006241255.1 | 2 - 3 | 179 | 102 | ||
GenBank mRNA | CB751206.1 | - | 1 - 2 | 383 | 102 | |
FQ209824.1 | - | 2 - 3 | 168 | 102 | ||
FQ210560.1 | - | 1 - 2 | 52 | 102 | ||
FQ210688.1 | - | 1 - 2 | 52 | 102 | ||
FQ225388.1 | - | 1 - 2 | 52 | 102 | ||
FQ225437.1 | - | 1 - 2 | 52 | 102 | ||
FQ234943.1 | - | 1 - 2 | 52 | 102 | ||
M17335.1 | - | 1 - 2 | 236 | 102 | ||
X06107.1 | - | 1 - 2 | 314 | 102 | ||
X06108.1 | - | 2 - 3 | 128 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the M17335.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
