Assay Details
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082477.2 | NP_001075946.2 | 3 - 4 | 385 | 63 | |
NM_001082478.1 | NP_001075947.1 | 3 - 4 | 682 | 63 | ||
XM_006241191.2 | XP_006241253.1 | 4 - 5 | 526 | 63 | ||
XM_006241192.1 | XP_006241254.1 | 4 - 5 | 440 | 63 | ||
XM_006241194.2 | XP_006241256.1 | 3 - 4 | 564 | 63 | ||
GenBank mRNA | FQ210560.1 | - | 3 - 4 | 398 | 63 | |
FQ220385.1 | - | 3 - 4 | 706 | 63 | ||
M15480.1 | - | 3 - 4 | 429 | 63 | ||
X06107.1 | - | 3 - 4 | 660 | 63 | ||
X06108.1 | - | 4 - 5 | 474 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
