Assay Details
Gene Symbol: Mettl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108089.1 | NP_001101559.1 | 8 - 9 | 1520 | 150 | |
GenBank mRNA | BC127512.1 | - | 9 - 10 | 1723 | 150 | |
BC161980.1 | - | 8 - 9 | 1522 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
