Assay Details
Gene Symbol: Slc5a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191987.1 | NP_001178916.1 | 2 - 3 | 412 | 137 | |
XM_006241232.2 | XP_006241294.1 | 2 - 3 | 2070 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
