Assay Details
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241616.1 | XP_006241678.1 | 3 - 4 | 394 | 61 | |
XM_006241617.1 | XP_006241679.1 | 3 - 4 | 394 | 61 | ||
XM_006241618.2 | XP_006241680.1 | 2 - 3 | 554 | 61 | ||
XM_008765476.1 | XP_008763698.1 | 2 - 3 | 335 | 61 | ||
XM_008765477.1 | XP_008763699.1 | 2 - 3 | 338 | 61 | ||
GenBank mRNA | FQ211706.1 | - | 3 - 4 | 394 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
