Assay Details
Gene Symbol: Eif3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004283.1 | NP_001004283.1 | 11 - 12 | 1165 | 128 | |
XM_006241938.2 | XP_006242000.1 | 11 - 12 | 1279 | 128 | ||
GenBank mRNA | BC079005.1 | - | 11 - 12 | 1165 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
