Assay Details
Gene Symbol: Eif3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004283.1 | NP_001004283.1 | 12 - 13 | 1293 | 145 | |
XM_006241938.2 | XP_006242000.1 | 12 - 13 | 1407 | 145 | ||
GenBank mRNA | BC079005.1 | - | 12 - 13 | 1293 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
