Assay Details
Gene Symbol: Rhpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241900.2 | XP_006241962.1 | 12 - 13 | 1649 | 72 | |
XM_006241901.2 | XP_006241963.1 | 12 - 13 | 1650 | 72 | ||
XM_006241902.1 | XP_006241964.1 | 10 - 11 | 1263 | 72 | ||
XM_008765628.1 | XP_008763850.1 | 12 - 13 | 3038 | 72 | ||
XM_216954.8 | XP_216954.4 | 12 - 13 | 1645 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
