Assay Details
Gene Symbol: Nipal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130559.1 | NP_001124031.1 | 10 - 11 | 1537 | 152 | |
XM_006241544.2 | XP_006241606.1 | 8 - 9 | 1144 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P108003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129765 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P108003 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P108003 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P108003 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P108003 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P274733 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10903239 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903242 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903238 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903245 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903235 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903241 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903244 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903240 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903236 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903243 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903237 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10903234 | - | - |
Rat Genome 230 2.0 Array | 1390474_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390474_at | - | - |
Affymetrix HT Rat Focus Array | 1390474_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390474_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700022923.rn.1 | TC0700002962.rn.1 | RTA TC |
Rat Expression Set 230 | 1390474_at | - | - |
Rat Expression Set 230 | 1390474_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903234 | - | - |
Rat Genome U34 Set | rc_AA859508_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20707937 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366328 | - | - |
