Assay Details
Gene Symbol: Slc25a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126269.1 | NP_001119741.1 | 5 - 6 | 576 | 89 | |
GenBank mRNA | BC166457.1 | - | 5 - 6 | 471 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
