Assay Details
Gene Symbol: Slc25a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126269.1 | NP_001119741.1 | 7 - 8 | 820 | 66 | |
GenBank mRNA | BC166457.1 | - | 7 - 8 | 715 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
