Assay Details
Gene Symbol: C1qtnf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034932.1 | NP_001030104.1 | 1 - 2 | 74 | 129 | |
XM_006241985.2 | XP_006242047.1 | 1 - 2 | 128 | 129 | ||
GenBank mRNA | BC101907.1 | - | 1 - 2 | 74 | 129 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC101907.1, NM_001034932.1, XM_006241985.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
