Assay Details
Gene Symbol: Tab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109976.2 | NP_001103446.1 | 2 - 3 | 181 | 62 | |
GenBank mRNA | CB605878.1 | - | 1 - 2 | 119 | 62 | |
CB779067.1 | - | 2 - 3 | 154 | 62 | ||
CB802111.1 | - | 2 - 3 | 240 | 62 | ||
FQ211519.1 | - | 2 - 3 | 616 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
