Assay Details
Gene Symbol: Tab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109976.2 | NP_001103446.1 | 4 - 5 | 423 | 72 | |
GenBank mRNA | CB605878.1 | - | 3 - 4 | 361 | 72 | |
EV771639.1 | - | 2 - 3 | 194 | 72 | ||
FQ211519.1 | - | 4 - 5 | 858 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
