Assay Details
Gene Symbol: Tab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109976.2 | NP_001103446.1 | 8 - 9 | 940 | 112 | |
GenBank mRNA | DY311072.1 | - | 3 - 4 | 284 | 112 | |
FQ211519.1 | - | 8 - 9 | 1375 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
