Assay Details
Gene Symbol: Poldip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130506.1 | NP_001123978.1 | 5 - 6 | 903 | 90 | |
XM_006242090.2 | XP_006242152.1 | 4 - 5 | 838 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
