Assay Details
Gene Symbol: Poldip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130506.1 | NP_001123978.1 | 7 - 8 | 1115 | 80 | |
XM_006242090.2 | XP_006242152.1 | 6 - 7 | 1050 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
