Assay Details
Gene Symbol: Angpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053546.1 | NP_445998.1 | 2 - 3 | 745 | 65 | |
XM_006241609.2 | XP_006241671.1 | 2 - 3 | 1087 | 65 | ||
XM_008765463.1 | XP_008763685.1 | 2 - 3 | 271 | 65 | ||
XM_008765464.1 | XP_008763686.1 | 2 - 3 | 2735 | 65 | ||
GenBank mRNA | AB080023.1 | - | 2 - 3 | 745 | 65 | |
AF030376.1 | - | 2 - 3 | 184 | 65 | ||
AF311727.1 | - | 2 - 3 | 184 | 65 | ||
AY052399.1 | - | 1 - 2 | 162 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
