Assay Details
Gene Symbol: Nhp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212515.2 | NP_997680.1 | 1 - 2 | 117 | 82 | |
GenBank mRNA | BC058493.1 | - | 1 - 2 | 120 | 82 | |
FQ217415.1 | - | 104 | 82 | |||
FQ220734.1 | - | 118 | 82 | |||
FQ233358.1 | - | 118 | 82 |
Gene Symbol: LOC100359574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760591.1 | XP_008758813.1 | 1 | 810 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC058493.1, NM_212515.2, XM_008760591.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
