Assay Details
Gene Symbol: Card10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130554.1 | NP_001124026.1 | 1 - 2 | 48 | 126 | |
XM_006241986.2 | XP_006242048.1 | 1 - 2 | 174 | 126 | ||
XM_006241987.2 | XP_006242049.1 | 1 - 2 | 174 | 126 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001130554.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
