Assay Details
Gene Symbol: Polr3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127295.2 | NP_001120767.1 | 4 - 5 | 335 | 121 | |
GenBank mRNA | BC167088.1 | - | 4 - 5 | 335 | 121 | |
FQ220736.1 | - | 4 - 5 | 369 | 121 | ||
FQ225628.1 | - | 4 - 5 | 335 | 121 | ||
FQ229831.1 | - | 4 - 5 | 370 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
