Assay Details
Gene Symbol: Sqle | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017136.2 | NP_058832.1 | 8 - 9 | 1412 | 151 | |
GenBank mRNA | BC097330.1 | - | 8 - 9 | 1412 | 151 | |
D37920.1 | - | 8 - 9 | 1543 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
