Assay Details
Gene Symbol: Slc39a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077669.1 | NP_001071137.1 | 6 - 7 | 1262 | 67 | |
XM_008765563.1 | XP_008763785.1 | 7 - 8 | 3439 | 67 | ||
XM_008765564.1 | XP_008763786.1 | 7 - 8 | 3317 | 67 | ||
GenBank mRNA | BC127514.1 | - | 6 - 7 | 1262 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
