Assay Details
Gene Symbol: Parvb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134780.1 | NP_001128252.1 | 2 - 3 | 231 | 112 | |
XM_006241404.2 | XP_006241466.1 | 2 - 3 | 304 | 112 | ||
XM_006241405.2 | XP_006241467.1 | 2 - 3 | 173 | 112 | ||
XM_006241406.2 | XP_006241468.1 | 2 - 3 | 1953 | 112 | ||
XM_006241407.2 | XP_006241469.1 | 2 - 3 | 409 | 112 | ||
XM_006241409.2 | XP_006241471.1 | 2 - 3 | 147 | 112 | ||
GenBank mRNA | BC166826.1 | - | 2 - 3 | 133 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
