Assay Details
Gene Symbol: Eif3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198751.1 | NP_942046.1 | 6 - 7 | 994 | 71 | |
GenBank mRNA | BC060586.1 | - | 6 - 7 | 994 | 71 | |
FQ216838.1 | - | 6 - 7 | 991 | 71 | ||
FQ220633.1 | - | 6 - 7 | 993 | 71 | ||
FQ228544.1 | - | 6 - 7 | 977 | 71 | ||
FQ229203.1 | - | 6 - 7 | 987 | 71 | ||
FQ232428.1 | - | 6 - 7 | 992 | 71 |
Gene Symbol: LOC100911110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750101.3 | XP_003750149.1 | 7 - 8 | 994 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P237915 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P072407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7788 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P237915 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P072407 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P072407 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P237915 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P072407 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P237915 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P237915 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74638 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90251 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43888 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57121 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6348 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32525 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10882731 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903687 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903685 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903688 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882732 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903686 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10882730 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10903684 | - | - |
Rat Genome 230 2.0 Array | 1397466_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388390_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397466_at | - | - |
Rat Genome 230 2.0 Array | 1388390_at | - | - |
Affymetrix HT Rat Focus Array | 1388390_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1397466_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388390_at | - | - |
Affymetrix HT Rat Focus Array | 1397466_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600000817.rn.1 | TC0600000140.rn.1 | RTA TC |
Rat Expression Set 230 | 1388390_PM_at | - | - |
Rat Expression Set 230 | 1388390_at | - | - |
Rat Expression Set 230 | 1397466_at | - | - |
Rat Expression Set 230 | 1397466_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903684 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10882730 | - | - |
Rat Genome U34 Set | rc_AI179125_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20895269 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355332 | - | - |
RatRef-12 Expression BeadChip | ILMN_51491 | - | - |
