Assay Details
Gene Symbol: Eif3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198751.1 | NP_942046.1 | 3 - 4 | 585 | 106 | |
GenBank mRNA | BC060586.1 | - | 3 - 4 | 585 | 106 | |
FQ216838.1 | - | 3 - 4 | 582 | 106 | ||
FQ220633.1 | - | 3 - 4 | 584 | 106 | ||
FQ228544.1 | - | 3 - 4 | 568 | 106 | ||
FQ229203.1 | - | 3 - 4 | 578 | 106 | ||
FQ232428.1 | - | 3 - 4 | 583 | 106 |
Gene Symbol: LOC100911110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750101.3 | XP_003750149.1 | 4 - 5 | 585 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
