Assay Details
Gene Symbol: Eif3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198751.1 | NP_942046.1 | 4 - 5 | 737 | 76 | |
GenBank mRNA | BC060586.1 | - | 4 - 5 | 737 | 76 | |
FQ216838.1 | - | 4 - 5 | 734 | 76 | ||
FQ220633.1 | - | 4 - 5 | 736 | 76 | ||
FQ228544.1 | - | 4 - 5 | 720 | 76 | ||
FQ229203.1 | - | 4 - 5 | 730 | 76 | ||
FQ232428.1 | - | 4 - 5 | 735 | 76 |
Gene Symbol: LOC100911110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750101.3 | XP_003750149.1 | 5 - 6 | 737 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
