Assay Details
Gene Symbol: Ttll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012200.1 | NP_001012200.1 | 4 - 5 | 372 | 76 | |
XM_006242102.2 | XP_006242164.1 | 4 - 5 | 377 | 76 | ||
XM_006242103.2 | XP_006242165.1 | 4 - 5 | 423 | 76 | ||
XM_006242104.1 | XP_006242166.1 | 3 - 4 | 418 | 76 | ||
XM_008765764.1 | XP_008763986.1 | 3 - 4 | 329 | 76 | ||
GenBank mRNA | BC087669.1 | - | 4 - 5 | 372 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
