Assay Details
Gene Symbol: Pacsin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130740.2 | NP_570096.2 | 5 - 6 | 823 | 84 | |
XM_006242049.2 | XP_006242111.1 | 4 - 5 | 1041 | 84 | ||
XM_006242050.2 | XP_006242112.1 | 5 - 6 | 1516 | 84 | ||
XM_006242051.1 | XP_006242113.1 | 5 - 6 | 765 | 84 | ||
XM_006242052.2 | XP_006242114.1 | 5 - 6 | 1426 | 84 | ||
XM_006242053.1 | XP_006242115.1 | 5 - 6 | 788 | 84 | ||
XM_006242054.2 | XP_006242116.1 | 4 - 5 | 1041 | 84 | ||
XM_006242055.1 | XP_006242117.1 | 5 - 6 | 765 | 84 | ||
XM_006242057.2 | XP_006242119.1 | 5 - 6 | 1517 | 84 | ||
XM_006242058.1 | XP_006242120.1 | 5 - 6 | 765 | 84 | ||
XM_006242059.2 | XP_006242121.1 | 5 - 6 | 1516 | 84 | ||
XM_006242060.1 | XP_006242122.1 | 5 - 6 | 765 | 84 | ||
XM_008765660.1 | XP_008763882.1 | 4 - 5 | 5264 | 84 | ||
XM_008765661.1 | XP_008763883.1 | 4 - 5 | 1383 | 84 | ||
XM_008765662.1 | XP_008763884.1 | 4 - 5 | 623 | 84 | ||
GenBank mRNA | AF139492.1 | - | 5 - 6 | 741 | 84 | |
AF139493.1 | - | 5 - 6 | 741 | 84 | ||
AF139494.1 | - | 5 - 6 | 741 | 84 | ||
AF139495.1 | - | 5 - 6 | 741 | 84 | ||
BC070911.1 | - | 5 - 6 | 823 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
