Assay Details
Gene Symbol: Gtse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130500.1 | NP_001123972.1 | 9 - 10 | 1985 | 85 | |
XM_006242160.2 | XP_006242222.1 | 10 - 11 | 2089 | 85 | ||
GenBank mRNA | FQ218328.1 | - | 10 - 11 | 2057 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
