Assay Details
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011991.1 | NP_001011991.1 | 1 - 2 | 124 | 77 | |
GenBank mRNA | AY500369.1 | - | 1 - 2 | 86 | 77 | |
BC081898.1 | - | 1 - 2 | 124 | 77 | ||
FQ212154.1 | - | 1 - 2 | 136 | 77 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY500369.1, BC081898.1, NM_001011991.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
