Assay Details
Gene Symbol: Ndrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011991.1 | NP_001011991.1 | 8 - 9 | 674 | 142 | |
XM_006241708.2 | XP_006241770.1 | 8 - 9 | 946 | 142 | ||
GenBank mRNA | AY500369.1 | - | 8 - 9 | 636 | 142 | |
BC081898.1 | - | 8 - 9 | 674 | 142 | ||
FQ212154.1 | - | 8 - 9 | 686 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
