Assay Details
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013104.1 | NP_001013122.1 | 3 - 4 | 1294 | 161 | |
XM_008765554.1 | XP_008763776.1 | 3 - 4 | 1422 | 161 | ||
XM_008765557.1 | XP_008763779.1 | 3 - 4 | 298 | 161 | ||
XM_008765558.1 | XP_008763780.1 | 3 - 4 | 305 | 161 | ||
XM_008765559.1 | XP_008763781.1 | 3 - 4 | 285 | 161 | ||
GenBank mRNA | BC079391.1 | - | 3 - 4 | 1294 | 161 | |
FQ220813.1 | - | 3 - 4 | 255 | 161 | ||
FQ225933.1 | - | 3 - 4 | 290 | 161 | ||
FQ227443.1 | - | 3 - 4 | 266 | 161 | ||
FQ233760.1 | - | 3 - 4 | 246 | 161 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
