Assay Details
Gene Symbol: Emc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113785.1 | NP_001107257.1 | 335 | 97 | ||
GenBank mRNA | BC158805.1 | - | 335 | 97 | ||
FQ214096.1 | - | 218 | 97 | |||
FQ215024.1 | - | 324 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
