Assay Details
Gene Symbol: Phkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080584.4 | NP_542151.1 | 7 - 8 | 1144 | 74 | |
XM_008759855.1 | XP_008758077.1 | 8 - 9 | 967 | 74 | ||
XM_008759856.1 | XP_008758078.1 | 8 - 9 | 1085 | 74 | ||
GenBank mRNA | BC107906.1 | - | 7 - 8 | 1004 | 74 | |
M73808.1 | - | 7 - 8 | 1144 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
