Assay Details
Gene Symbol: Phkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080584.4 | NP_542151.1 | 2 - 3 | 612 | 81 | |
XM_008759855.1 | XP_008758077.1 | 3 - 4 | 435 | 81 | ||
XM_008759856.1 | XP_008758078.1 | 3 - 4 | 553 | 81 | ||
GenBank mRNA | BC128715.1 | - | 2 - 3 | 541 | 81 | |
M73808.1 | - | 2 - 3 | 612 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
