Assay Details
Gene Symbol: Igsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133542.2 | NP_598226.1 | 5 - 6 | 702 | 134 | |
GenBank mRNA | AJ223184.1 | - | 5 - 6 | 702 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
