Assay Details
Gene Symbol: Nars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034921.1 | NP_001030093.1 | 2 - 3 | 493 | 71 | |
GenBank mRNA | BC104691.1 | - | 2 - 3 | 493 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
