Assay Details
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106282.1 | NP_001099752.1 | 1 - 2 | 103 | 76 | |
GenBank mRNA | FQ212514.1 | - | 1 - 2 | 300 | 76 | |
FQ214311.1 | - | 1 - 2 | 152 | 76 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001106282.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
