Assay Details
Gene Symbol: Ctsc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017097.1 | NP_058793.1 | 3 - 4 | 532 | 130 | |
GenBank mRNA | D90404.1 | - | 3 - 4 | 532 | 130 | |
FQ218311.1 | - | 592 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
