Assay Details
Gene Symbol: Slc5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022590.2 | NP_072112.2 | 7 - 8 | 886 | 88 | |
GenBank mRNA | AB070850.1 | - | 6 - 7 | 1021 | 88 | |
AB070976.1 | - | 7 - 8 | 881 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
