Assay Details
Gene Symbol: Ppp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134359.1 | NP_599186.1 | 2 - 3 | 324 | 92 | |
XM_006230206.1 | XP_006230268.1 | 3 - 4 | 320 | 92 | ||
GenBank mRNA | BC091574.1 | - | 2 - 3 | 324 | 92 | |
FQ216602.1 | - | 3 - 4 | 280 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
