Assay Details
Gene Symbol: Ampd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031544.1 | NP_113732.1 | 11 - 12 | 1841 | 106 | |
XM_006230010.2 | XP_006230072.1 | 11 - 12 | 2609 | 106 | ||
XM_006230011.2 | XP_006230073.1 | 11 - 12 | 1974 | 106 | ||
XM_006230012.2 | XP_006230074.1 | 11 - 12 | 2444 | 106 | ||
XM_006230013.2 | XP_006230075.1 | 11 - 12 | 2456 | 106 | ||
XM_006230014.2 | XP_006230076.1 | 11 - 12 | 2360 | 106 | ||
XM_006230015.2 | XP_006230077.1 | 11 - 12 | 2588 | 106 | ||
XM_008759688.1 | XP_008757910.1 | 11 - 12 | 1851 | 106 | ||
GenBank mRNA | U90888.1 | - | 11 - 12 | 1841 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
