Assay Details
Gene Symbol: Ino80e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013900.1 | NP_001013922.1 | 2 - 3 | 272 | 110 | |
XM_006230244.1 | XP_006230306.1 | 2 - 3 | 198 | 110 | ||
XM_006230245.1 | XP_006230307.1 | 2 - 3 | 198 | 110 | ||
GenBank mRNA | BC079045.1 | - | 2 - 3 | 272 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
