Assay Details
Gene Symbol: Klhl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229818.1 | XP_006229880.1 | 5 - 6 | 5462 | 95 | |
XM_218953.6 | XP_218953.4 | 4 - 5 | 1462 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
