Assay Details
Gene Symbol: Gdpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728806.4 | XP_002728852.3 | 6 - 7 | 1159 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
