Assay Details
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106295.1 | NP_001099765.1 | 8 - 9 | 1402 | 133 | |
XM_006230224.1 | XP_006230286.1 | 8 - 9 | 1313 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
